siRNA Details
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053819.1 | NP_446271.1 | 4, 5 | 372 | ||
XM_006256608.2 | XP_006256670.1 | 4, 5 | 436 | |||
XM_008773034.1 | XP_008771256.1 | 3, 4 | 346 | |||
GenBank mRNA | AF411319.1 | - | 1, 2 | 122 | ||
AY550026.1 | - | 3, 4 | 361 | |||
BC099821.1 | - | 4, 5 | 370 | 7930800 | ||
FQ219856.1 | - | 4, 5 | 382 | |||
FQ221763.1 | - | 4, 5 | 382 | |||
FQ222271.1 | - | 4, 5 | 382 | |||
FQ222437.1 | - | 4, 5 | 385 | |||
FQ222915.1 | - | 4, 5 | 382 | |||
FQ228400.1 | - | 4, 5 | 381 | |||
FQ230039.1 | - | 4, 5 | 379 | |||
L31883.1 | - | 4, 5 | 372 | |||
U06179.1 | - | 3, 4 | 355 | |||
U16022.1 | - | 2, 3 | 266 |
