siRNA Details
| Gene Symbol: Pou2f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105745.1 | NP_001099215.1 | 5 | 381 | ||
| GenBank mRNA | L23862.1 | - | 5 | 351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou2f3| Gene Symbol: Pou2f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105745.1 | NP_001099215.1 | 5 | 381 | ||
| GenBank mRNA | L23862.1 | - | 5 | 351 | ||