siRNA Details
| Gene Symbol: Pou6f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105746.1 | NP_001099216.1 | 2 | 863 | ||
| XM_006242298.2 | XP_006242360.1 | 8 | 1526 | |||
| XM_006242301.2 | XP_006242363.1 | 7 | 1763 | |||
| XM_008765654.1 | XP_008763876.1 | 7 | 1877 | |||
| XM_008765655.1 | XP_008763877.1 | 8 | 1538 | |||
| XM_008765656.1 | XP_008763878.1 | 8 | 1596 | |||
| XM_008765657.1 | XP_008763879.1 | 7 | 1874 | |||
| XM_008765658.1 | XP_008763880.1 | 7 | 1865 | |||
| GenBank mRNA | L23204.1 | - | 3 | 840 | ||

Pou6f1
Genomic Map