siRNA Details
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053821.2 | NP_446273.1 | 3 | 398 | ||
GenBank mRNA | BC072505.1 | - | 3 | 398 | 7099019 | |
L19699.1 | - | 2 | 246 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053821.2 | NP_446273.1 | 3 | 398 | ||
GenBank mRNA | BC072505.1 | - | 3 | 398 | 7099019 | |
L19699.1 | - | 2 | 246 |