siRNA Details
| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053823.1 | NP_446275.1 | 6 | 707 | ||
| GenBank mRNA | L14933.2 | - | 6 | 708 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk5| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053823.1 | NP_446275.1 | 6 | 707 | ||
| GenBank mRNA | L14933.2 | - | 6 | 708 | ||