siRNA Details
| Gene Symbol: Csnk2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053824.2 | NP_446276.1 | 7 | 593 | ||
| XM_006235220.2 | XP_006235282.1 | 6 | 653 | |||
| XM_006235221.2 | XP_006235283.1 | 6 | 694 | |||
| GenBank mRNA | BC091130.1 | - | 7 | 593 | 7367296 | |
| L15618.1 | - | 1210 | ||||

Csnk2a1
Genomic Map