siRNA Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB782828.1 | - | 3 | 283 | ||
| L27129.1 | - | 3 | 332 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016700, BC053027, AK030767, AK043102, AK139498, and others.

Mapk8
Genomic Map