siRNA Details
| Gene Symbol: Nup107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053830.2 | NP_446282.2 | 16 | 1515 | ||
| GenBank mRNA | EV764995.1 | - | 462 | |||
| L31840.1 | - | 16 | 1535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup107| Gene Symbol: Nup107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053830.2 | NP_446282.2 | 16 | 1515 | ||
| GenBank mRNA | EV764995.1 | - | 462 | |||
| L31840.1 | - | 16 | 1535 | |||