siRNA Details
| Gene Symbol: Kcnj9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053834.1 | NP_446286.1 | 3 | 408 | ||
| XM_008769730.1 | XP_008767952.1 | 1 | 1370 | |||
| GenBank mRNA | L77929.1 | - | 3 | 408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj9| Gene Symbol: Kcnj9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053834.1 | NP_446286.1 | 3 | 408 | ||
| XM_008769730.1 | XP_008767952.1 | 1 | 1370 | |||
| GenBank mRNA | L77929.1 | - | 3 | 408 | ||