siRNA Details
| Gene Symbol: Cltb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053835.1 | NP_446287.1 | 6 | 561 | ||
| XM_006253595.2 | XP_006253657.1 | 5 | 595 | |||
| GenBank mRNA | FQ213983.1 | - | 6 | 651 | ||
| FQ229322.1 | - | 5 | 525 | |||
| M15883.1 | - | 6 | 561 | |||
| M19262.1 | - | 5 | 507 | |||

Cltb
Genomic Map