siRNA Details
| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169113.1 | NP_001162584.1 | 2 | 373 | ||
| GenBank mRNA | BC082020.1 | - | 2 | 341 | ||
| BC098947.1 | - | 2 | 338 | |||
| FM055449.1 | - | 2 | 375 | |||
| M31051.1 | - | 2 | 229 | |||
| Z11994.1 | - | 333 | ||||

Lrpap1
Genomic Map