siRNA Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053851.1 | NP_446303.1 | 10, 11 | 1424 | ||
| XM_006254298.2 | XP_006254360.1 | 11, 12 | 1392 | |||
| XM_006254300.2 | XP_006254362.1 | 10, 11 | 1333 | |||
| XM_006254301.2 | XP_006254363.1 | 10, 11 | 2290 | |||
| XM_006254303.2 | XP_006254365.1 | 10, 11 | 1589 | |||
| XM_006254304.2 | XP_006254366.1 | 10, 11 | 2241 | |||
| GenBank mRNA | AF394941.1 | - | 10, 11 | 1315 | ||
| AF423193.1 | - | 10, 11 | 1247 | |||
| AY190119.1 | - | 11, 12 | 1130 | |||
| AY190120.1 | - | 10, 11 | 1070 | |||
| M80545.1 | - | 10, 11 | 1424 | |||

Cacnb2
Genomic Map