siRNA Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053851.1 | NP_446303.1 | 5, 6 | 883 | ||
| XM_006254298.2 | XP_006254360.1 | 6, 7 | 851 | |||
| XM_006254300.2 | XP_006254362.1 | 5, 6 | 792 | |||
| XM_006254301.2 | XP_006254363.1 | 5, 6 | 1749 | |||
| XM_006254303.2 | XP_006254365.1 | 5, 6 | 1048 | |||
| GenBank mRNA | AF394941.1 | - | 5, 6 | 774 | ||
| AF423193.1 | - | 5, 6 | 706 | |||
| AY190119.1 | - | 6, 7 | 589 | |||
| AY190120.1 | - | 5, 6 | 529 | |||
| M80545.1 | - | 5, 6 | 883 | |||

Cacnb2
Genomic Map