siRNA Details
Gene Symbol: Atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058213.1 | NP_478120.1 | 6 | 654 | ||
GenBank mRNA | BC168245.1 | - | 6 | 667 | 7935496 | |
M99223.1 | - | 6 | 654 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058213.1 | NP_478120.1 | 6 | 654 | ||
GenBank mRNA | BC168245.1 | - | 6 | 667 | 7935496 | |
M99223.1 | - | 6 | 654 |