siRNA Details
| Gene Symbol: Eif4ebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053857.2 | NP_446309.1 | 2 | 353 | ||
| GenBank mRNA | U05014.1 | - | 2 | 353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif4ebp1| Gene Symbol: Eif4ebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053857.2 | NP_446309.1 | 2 | 353 | ||
| GenBank mRNA | U05014.1 | - | 2 | 353 | ||