siRNA Details
| Gene Symbol: Lgals8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053862.2 | NP_446314.2 | 8 | 737 | ||
| XM_006254166.2 | XP_006254228.1 | 8 | 1003 | |||
| XM_006254167.2 | XP_006254229.1 | 8 | 749 | |||
| XM_006254168.1 | XP_006254230.1 | 8 | 776 | |||
| GenBank mRNA | BC072488.1 | - | 8 | 737 | 7104910 | |
| U09824.1 | - | 7 | 717 | |||

Lgals8
Genomic Map