siRNA Details
| Gene Symbol: Lgals8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053862.2 | NP_446314.2 | 5, 6 | 600 | ||
| XM_006254166.2 | XP_006254228.1 | 5, 6 | 866 | |||
| XM_006254167.2 | XP_006254229.1 | 5, 6 | 612 | |||
| XM_006254168.1 | XP_006254230.1 | 5, 6 | 639 | |||
| GenBank mRNA | BC072488.1 | - | 5, 6 | 600 | 7104910 | |
| U09824.1 | - | 4, 5 | 580 | |||

Lgals8
Genomic Map