siRNA Details
| Gene Symbol: Slc28a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053863.1 | NP_446315.1 | 14 | 1639 | ||
| XM_006229422.2 | XP_006229484.1 | 16 | 2664 | |||
| XM_006229423.2 | XP_006229485.1 | 16 | 2776 | |||
| GenBank mRNA | U10279.1 | - | 14 | 1639 | ||

Slc28a1
Genomic Map