siRNA Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053865.1 | NP_446317.1 | 2 | 987 | ||
| GenBank mRNA | U17604.1 | - | 2 | 987 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtn1| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053865.1 | NP_446317.1 | 2 | 987 | ||
| GenBank mRNA | U17604.1 | - | 2 | 987 | ||