siRNA Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053875.1 | NP_446327.1 | 13 | 1965 | ||
| XM_008762757.1 | XP_008760979.1 | 13 | 2060 | |||
| GenBank mRNA | U32498.1 | - | 13 | 1965 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exoc4| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053875.1 | NP_446327.1 | 13 | 1965 | ||
| XM_008762757.1 | XP_008760979.1 | 13 | 2060 | |||
| GenBank mRNA | U32498.1 | - | 13 | 1965 | ||