siRNA Details
| Gene Symbol: Cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053879.1 | NP_446331.1 | 24 | 3269 | ||
| GenBank mRNA | U35371.1 | - | 24 | 3269 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn4| Gene Symbol: Cntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053879.1 | NP_446331.1 | 24 | 3269 | ||
| GenBank mRNA | U35371.1 | - | 24 | 3269 | ||