siRNA Details
| Gene Symbol: Map3k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053887.1 | NP_446339.1 | 9 | 2140 | ||
| GenBank mRNA | JQ013733.1 | - | 9 | 1657 | ||
| U48596.1 | - | 9 | 2140 | |||
| Gene Symbol: LOC100912399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231926.2 | XP_006231988.2 | 2121 | |||

Map3k1
Genomic Map