siRNA Details
Gene Symbol: Vwf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053889.1 | NP_446341.1 | 47 | 8009 | ||
XM_008763279.1 | XP_008761501.1 | 47 | 7998 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
