siRNA Details
| Gene Symbol: Sdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053893.3 | NP_446345.3 | 4 | 1184 | ||
| GenBank mRNA | U52825.1 | - | 4 | 1187 | ||
| U73184.1 | - | 4 | 1252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdc3| Gene Symbol: Sdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053893.3 | NP_446345.3 | 4 | 1184 | ||
| GenBank mRNA | U52825.1 | - | 4 | 1187 | ||
| U73184.1 | - | 4 | 1252 | |||