siRNA Details
| Gene Symbol: Pgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229825.2 | XP_006229887.1 | 8 | 1763 | ||
| XM_006229830.2 | XP_006229892.1 | 8 | 1752 | |||
| XM_006229831.2 | XP_006229893.1 | 8 | 1575 | |||
| XM_006229832.2 | XP_006229894.1 | 8 | 1563 | |||
| XM_006229834.2 | XP_006229896.1 | 7 | 1531 | |||
| XM_006229835.2 | XP_006229897.1 | 5 | 1914 | |||
| XM_006229837.2 | XP_006229899.1 | 7 | 1346 | |||
| XM_006229838.2 | XP_006229900.1 | 7 | 1334 | |||
| XM_008759662.1 | XP_008757884.1 | 5 | 1314 | |||
| XM_008759663.1 | XP_008757885.1 | 5 | 1855 | |||
| GenBank mRNA | AB236144.1 | - | 6 | 821 | ||
| FQ230423.1 | - | 1 | 50 | |||
| U57715.1 | - | 7 | 1326 | |||

Pgap2
Genomic Map