siRNA Details
| Gene Symbol: Ptpru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191575.1 | NP_001178504.1 | 13 | 2294 | ||
| XM_006238997.2 | XP_006239059.1 | 13 | 2310 | |||
| XM_006238998.2 | XP_006239060.1 | 13 | 2310 | |||
| XM_008764130.1 | XP_008762352.1 | 13 | 2311 | |||
| GenBank mRNA | U66566.1 | - | 3 | 355 | ||

Ptpru
Genomic Map