siRNA Details
| Gene Symbol: Ptpru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191575.1 | NP_001178504.1 | 30 | 4325 | ||
| XM_006238997.2 | XP_006239059.1 | 29 | 4311 | |||
| XM_006238998.2 | XP_006239060.1 | 29 | 4302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpru| Gene Symbol: Ptpru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191575.1 | NP_001178504.1 | 30 | 4325 | ||
| XM_006238997.2 | XP_006239059.1 | 29 | 4311 | |||
| XM_006238998.2 | XP_006239060.1 | 29 | 4302 | |||