siRNA Details
| Gene Symbol: Dlgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053901.1 | NP_446353.2 | 9 | 1998 | ||
| GenBank mRNA | U67138.1 | - | 9 | 1998 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlgap2| Gene Symbol: Dlgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053901.1 | NP_446353.2 | 9 | 1998 | ||
| GenBank mRNA | U67138.1 | - | 9 | 1998 | ||