siRNA Details
Gene Symbol: Dlgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053901.1 | NP_446353.2 | 6 | 1305 | ||
GenBank mRNA | U67138.1 | - | 6 | 1305 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dlgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053901.1 | NP_446353.2 | 6 | 1305 | ||
GenBank mRNA | U67138.1 | - | 6 | 1305 |