siRNA Details
| Gene Symbol: Kynu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053902.2 | NP_446354.1 | 3 | 282 | ||
| XM_006234138.2 | XP_006234200.1 | 3 | 345 | |||
| XM_008761695.1 | XP_008759917.1 | 3 | 345 | |||
| GenBank mRNA | BC078762.1 | - | 3 | 265 | 7128911 | |
| U68168.1 | - | 3 | 283 | |||

Kynu
Genomic Map