siRNA Details
| Gene Symbol: Lmnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053905.1 | NP_446357.1 | 7 | 1686 | ||
| GenBank mRNA | U72353.1 | - | 7 | 1686 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmnb1| Gene Symbol: Lmnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053905.1 | NP_446357.1 | 7 | 1686 | ||
| GenBank mRNA | U72353.1 | - | 7 | 1686 | ||