siRNA Details
| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053908.1 | NP_446360.1 | 10 | 1355 | ||
| GenBank mRNA | AF468653.1 | - | 3 | 324 | ||
| BC099824.1 | - | 10 | 1212 | 7932620 | ||
| FQ232747.1 | - | 4 | 404 | |||
| U77038.1 | - | 10 | 1355 | |||

Ptpn6
Genomic Map