siRNA Details
| Gene Symbol: Cyth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053910.1 | NP_446362.1 | 3, 4 | 238 | ||
| XM_006247784.2 | XP_006247846.1 | 3, 4 | 282 | |||
| XM_006247785.2 | XP_006247847.1 | 3, 4 | 1373 | |||
| XM_008768306.1 | XP_008766528.1 | 2, 3 | 2678 | |||
| XM_008768307.1 | XP_008766529.1 | 2, 3 | 235 | |||
| GenBank mRNA | BC166417.1 | - | 3, 4 | 238 | 9028626 | |
| U83895.1 | - | 3, 4 | 238 | |||

Cyth1
Genomic Map