siRNA Details
| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053916.1 | NP_446368.1 | 6 | 1226 | ||
| XM_006228108.2 | XP_006228170.1 | 6 | 1222 | |||
| GenBank mRNA | BC166843.1 | - | 6 | 1226 | 9028761 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim28| Gene Symbol: Trim28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053916.1 | NP_446368.1 | 6 | 1226 | ||
| XM_006228108.2 | XP_006228170.1 | 6 | 1222 | |||
| GenBank mRNA | BC166843.1 | - | 6 | 1226 | 9028761 | |