siRNA Details
| Gene Symbol: Trip10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053920.1 | NP_446372.1 | 5, 6 | 455 | ||
| GenBank mRNA | AB006914.1 | - | 5, 6 | 455 | ||
| BC061840.1 | - | 5, 6 | 457 | 5600051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trip10| Gene Symbol: Trip10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053920.1 | NP_446372.1 | 5, 6 | 455 | ||
| GenBank mRNA | AB006914.1 | - | 5, 6 | 455 | ||
| BC061840.1 | - | 5, 6 | 457 | 5600051 | ||