siRNA Details
| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053928.2 | NP_446380.1 | 3 | 652 | ||
| GenBank mRNA | AB032739.1 | - | 3 | 313 | ||
| BC090072.1 | - | 3 | 652 | 7113593 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2n| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053928.2 | NP_446380.1 | 3 | 652 | ||
| GenBank mRNA | AB032739.1 | - | 3 | 313 | ||
| BC090072.1 | - | 3 | 652 | 7113593 | ||