siRNA Details
| Gene Symbol: Gp1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053930.4 | NP_446382.1 | 2 | 868 | ||
| GenBank mRNA | AB027146.1 | - | 1 | 1564 | ||
| FQ227461.1 | - | 2 | 885 | |||
| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053931.3 | NP_446383.3 | 11 | 3570 | ||
| GenBank mRNA | AB027143.1 | - | 11 | 3571 | ||

Gp1bb
Genomic Map