siRNA Details
| Gene Symbol: Pcdha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053933.1 | NP_446385.1 | 1 | 627 | ||
| GenBank mRNA | AB113387.1 | - | 1 | 627 | ||
| AF177687.1 | - | 1 | 241 | |||
| AF539750.1 | - | 1 | 627 | |||
| AY573978.1 | - | 1 | 627 | |||
| BC085793.1 | - | 1 | 718 | 7109469 | ||

Pcdha4
Genomic Map