siRNA Details
| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053935.1 | NP_446387.1 | 4 | 288 | ||
| XM_006238359.2 | XP_006238421.1 | 4 | 1672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3gl2| Gene Symbol: Sh3gl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053935.1 | NP_446387.1 | 4 | 288 | ||
| XM_006238359.2 | XP_006238421.1 | 4 | 1672 | |||