siRNA Details
| Gene Symbol: Pcdha10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053939.1 | NP_446391.1 | 1 | 2061 | ||
| GenBank mRNA | AB113393.1 | - | 1 | 2061 | ||
| AY573972.1 | - | 1 | 2061 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha10| Gene Symbol: Pcdha10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053939.1 | NP_446391.1 | 1 | 2061 | ||
| GenBank mRNA | AB113393.1 | - | 1 | 2061 | ||
| AY573972.1 | - | 1 | 2061 | |||