siRNA Details
| Gene Symbol: Pcdha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053941.1 | NP_446393.1 | 1 | 1039 | ||
| GenBank mRNA | AB113386.1 | - | 1 | 1039 | ||
| AF539749.1 | - | 1 | 662 | |||
| AY573977.1 | - | 1 | 1039 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha3| Gene Symbol: Pcdha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053941.1 | NP_446393.1 | 1 | 1039 | ||
| GenBank mRNA | AB113386.1 | - | 1 | 1039 | ||
| AF539749.1 | - | 1 | 662 | |||
| AY573977.1 | - | 1 | 1039 | |||