siRNA Details
| Gene Symbol: Pcdha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053942.1 | NP_446394.1 | 1 | 1889 | ||
| GenBank mRNA | AB113391.1 | - | 1 | 1889 | ||
| AY573982.1 | - | 1 | 1889 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha8| Gene Symbol: Pcdha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053942.1 | NP_446394.1 | 1 | 1889 | ||
| GenBank mRNA | AB113391.1 | - | 1 | 1889 | ||
| AY573982.1 | - | 1 | 1889 | |||