siRNA Details
| Gene Symbol: Pcdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053944.1 | NP_446396.1 | 1 | 424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdh12| Gene Symbol: Pcdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053944.1 | NP_446396.1 | 1 | 424 | ||