siRNA Details
| Gene Symbol: Il16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105749.1 | NP_001099219.1 | 14, 15 | 3320 | ||
| XM_006229488.2 | XP_006229550.1 | 14, 15 | 4967 | |||
| XM_006229489.2 | XP_006229551.1 | 14, 15 | 4966 | |||
| XM_008759599.1 | XP_008757821.1 | 14, 15 | 6783 | |||
| XM_008759600.1 | XP_008757822.1 | 15, 16 | 3339 | |||
| XM_008759601.1 | XP_008757823.1 | 2, 3 | 1264 | |||

Il16
Genomic Map