siRNA Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053949.1 | NP_446401.1 | 8 | 2137 | ||
| XM_006235867.2 | XP_006235929.1 | 8 | 2270 | |||
| XM_008762630.1 | XP_008760852.1 | 4 | 1458 | |||
| GenBank mRNA | AY669863.1 | - | 4 | 1111 | ||
| Z96106.1 | - | 8 | 2137 | |||

Kcnh2
Genomic Map