siRNA Details
| Gene Symbol: Crym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053955.1 | NP_446407.1 | 3 | 371 | ||
| GenBank mRNA | BC088121.1 | - | 3 | 402 | 7323247 | |
| Y17328.1 | - | 3 | 371 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016669, AK143116, AK163801, BC045159, AK163781, and others.

Crym
Genomic Map