siRNA Details
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053955.1 | NP_446407.1 | 4, 5 | 526 | ||
GenBank mRNA | BC088121.1 | - | 4, 5 | 557 | 7323247 | |
Y17328.1 | - | 4, 5 | 526 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
