siRNA Details
| Gene Symbol: Rhbdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191822.1 | NP_001178751.1 | 8 | 1037 | ||
| XM_006245894.2 | XP_006245956.1 | 7 | 2617 | |||
| XM_008767537.1 | XP_008765759.1 | 8 | 2791 | |||
| GenBank mRNA | Y17258.1 | - | 5 | 421 | ||

Rhbdl1
Genomic Map