siRNA Details
| Gene Symbol: Rhbdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191822.1 | NP_001178751.1 | 3 | 357 | ||
| XM_006245894.2 | XP_006245956.1 | 2 | 1937 | |||
| XM_008767537.1 | XP_008765759.1 | 3 | 2111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdl1| Gene Symbol: Rhbdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191822.1 | NP_001178751.1 | 3 | 357 | ||
| XM_006245894.2 | XP_006245956.1 | 2 | 1937 | |||
| XM_008767537.1 | XP_008765759.1 | 3 | 2111 | |||