siRNA Details
Gene Symbol: Apbb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053957.1 | NP_446409.1 | 13 | 1735 | ||
XM_006254495.1 | XP_006254557.1 | 13 | 1736 | |||
GenBank mRNA | BC097935.1 | - | 10 | 1988 | ||
Y13413.1 | - | 13 | 1735 |
